Skip to Content
Merck
All Photos(1)

Documents

SML0264

Sigma-Aldrich

AEG 3482

≥98% (HPLC)

Synonym(s):

6-Phenyl-imidazo[2,1-b]-1,3,4-thiadiazole-2-sulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8N4O2S2
CAS Number:
Molecular Weight:
280.33
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

faintly yellow to dark yellow

solubility

DMSO: ≥15 mg/mL

storage temp.

2-8°C

SMILES string

NS(=O)(=O)c1nn2cc(nc2s1)-c3ccccc3

InChI

1S/C10H8N4O2S2/c11-18(15,16)10-13-14-6-8(12-9(14)17-10)7-4-2-1-3-5-7/h1-6H,(H2,11,15,16)

InChI key

MQUYTXDAVCOCMX-UHFFFAOYSA-N

Biochem/physiol Actions

AEG 3482 is an imidazothiadiazole sulfonamide of 281 D with an anti-apoptotic property.
AEG3284 is a compound that binds to heat shock protein 90 (HSP90), leading to heat shock factor 1 (HSF1) dependent expression of HSP70, an endogenous inhibitor of c-Jun N-terminal kinase (JNK) activity. Treatment of PC12 cells with AEF3284 blocks JNK dependent apoptosis.

Features and Benefits

This compound is featured on the MAPKs page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amir H Salehi et al.
Chemistry & biology, 13(2), 213-223 (2006-02-24)
We describe a group of small-molecule inhibitors of Jun kinase (JNK)-dependent apoptosis. AEG3482, the parental compound, was identified in a screening effort designed to detect compounds that reduce apoptosis of neonatal sympathetic neurons after NGF withdrawal. We show that AEG3482
Karthiga Santhana Kumar et al.
SpringerPlus, 4, 19-19 (2015-01-28)
Medulloblastoma (MB) comprises four molecularly and genetically distinct subgroups of embryonal brain tumors that develop in the cerebellum. MB mostly affects infants and children and is difficult to treat because of frequent dissemination of tumor cells within the leptomeningeal space.
Zhen Ning Wee et al.
Nature communications, 6, 8746-8746 (2015-10-28)
Metastatic tumour recurrence due to failed treatments remains a major challenge of breast cancer clinical management. Here we report that interleukin-1 receptor-associated kinase 1 (IRAK1) is overexpressed in a subset of breast cancers, in particular triple-negative breast cancer (TNBC), where

Articles

The mitogen-activated protein kinase (MAPK) family consists of stress-activated (SAPK) and MAPKs. Learn about their network of signal transduction cascades that mediate cellular responses to a diverse range of stimuli.

The mitogen-activated protein kinase (MAPK) family consists of stress-activated (SAPK) and MAPKs. Learn about their network of signal transduction cascades that mediate cellular responses to a diverse range of stimuli.

The mitogen-activated protein kinase (MAPK) family consists of stress-activated (SAPK) and MAPKs. Learn about their network of signal transduction cascades that mediate cellular responses to a diverse range of stimuli.

The mitogen-activated protein kinase (MAPK) family consists of stress-activated (SAPK) and MAPKs. Learn about their network of signal transduction cascades that mediate cellular responses to a diverse range of stimuli.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service