Skip to Content
Merck
All Photos(1)

Key Documents

EP5820754001

Rotor for Eppendorf® 5804/5804R & 5810/5810R Centrifuges

S-4-104, for 5810/5810R, aerosol-tight, with 4 x 750 mL plate buckets

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103909
NACRES:
NB.82

feature

to hold 4 x plate buckets (aerosol-tight, capable)
aerosol-tight
for 5810/5810R
high-capacity swing-bucket rotor
with 4 x 750 mL plate buckets
with 4 × DWP buckets with plate carriers

packaging

pack of 1 ea

manufacturer/tradename

Eppendorf® 5820754001 (Global)

parameter

2,568 × g max. RCF
3,900 rpm

technique(s)

centrifugation: suitable

compatibility

for use with centrifuge models 5804/5804R/5810/5810R

Looking for similar products? Visit Product Comparison Guide

General description

Medium capacity swing-bucket rotor designed for Eppendorf® 5804/5804R & 5810/5810R Centrifuges. The 4-place rotor can accommodate 4 deep well plate buckets of 750 mL.

Application

Rotor for Eppendorf® 5804/5804R & 5810/5810R centrifuges are suitable for blood, bacteria, cell culture, and plate centrifugation.

Features and Benefits

  • Medium capacity swing-bucket Rotor
  • aerosol-tight capable for safe centrifugation of hazardous samples
  • The rotor reaches 3,900 rpm maximum speed with 3,214 × g maximum RCF
  • Includes 4 deep well plates (DWP) buckets

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service