Skip to Content
Merck
All Photos(1)

Key Documents

EP5804740009

Eppendorf® Deepwell Plate Rotor A-2-DWP

incl. 2 buckets

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103909
NACRES:
NB.82

feature

to hold 2 x Deepwell plates (max height 89 mm)
to hold 2 x max height 89 mm
incl. 2 buckets

packaging

pack of 1 ea

manufacturer/tradename

Eppendorf® 5804740009

parameter

3,700 max. RCF (2,250 xg)

technique(s)

centrifugation: suitable

compatibility

for use with centrifuge models 5804/5804R/5810/5810R

Features and Benefits

  • Incl. 2 buckets
  • Stacked plates in various combinations can be effectively centrifuged
  • Max. loading height 89 mm
  • Simultaneous processing of up to 192 samples
  • Autoclavable at 121 °C for 20 mins
  • The rotor can speed up to a maximum of 2.250 x g (3.700 rpm)
  • Compatible with 5804/5804R and 5810/5810R centrifuge models

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service