371206
Dkk-1, His•Tag®, Mouse, Recombinant
Dkk-1, His•Tag, Mouse, Recombinant, consists of amino acids 30-272, fused at the C-terminus to a His•Tag sequence, and expressed in NS0-dervied mouse myeloma cells.
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
Recommended Products
recombinant
expressed in mouse NSO cells (mouse myeloma cells)
Quality Level
Assay
≥95% (SDS-PAGE)
form
lyophilized
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
impurities
≤1.0 EU/μg protein Endotoxin
shipped in
ambient
storage temp.
−20°C
General description
Dkk-1, His•Tag, Mouse, Recombinant, consists of amino acids 30-272, fused at the C-terminus to a His•Tag sequence, and expressed in NS0-dervied mouse myeloma cells.
Recombinant, murine Dickkopf related protein-1 (Dkk-1), consisting of amino acids 30-272, fused at the C-terminus to a His•Tag sequence, and expressed in NS0-dervied mouse myeloma cells. Dkk-1 is a well-documented antagonist of the Wnt signaling pathway. The balance between Dkk-1 inhibition and Wnt signaling is essential for bone formation and homeostasis.
Biochem/physiol Actions
ND₅₀: 0.2-1 µg/ml as measured by the inhibition of Wnt-3a-induced alkaline phosphatase production in MC3T3-E1 mouse osteoblastic cells.
Reduction of Axis-inducing activity: 60-100% reduction (using 1 ng rmDkk-1 per embryo) in the number of secondary axes produced in the presence of 10-20 pg Xwnt-8 mRNA using early Xenopus embryos.
Reduction of Axis-inducing activity: 60-100% reduction (using 1 ng rmDkk-1 per embryo) in the number of secondary axes produced in the presence of 10-20 pg Xwnt-8 mRNA using early Xenopus embryos.
Warning
Toxicity: Regulatory Review (Z)
Physical form
Lyophilized from 0.2 µm filtered PBS, BSA.
Reconstitution
Following reconstitution refrigerate (4°C) for short-term storage or aliquot and freeze (-20°C) for long-term storage. Stock solutions are stable for up to 1 month at 4°C or for up to 3 months at -20°C.
Reconstitute to 100 µg/ml with sterile PBS containing ≥1% human or bovine serum albumin.
Other Notes
Pinzone, J.J. et al. 2009. Blood113, 517.
Li, J. et al. 2006. Bone36, 754.
Morvan, F. et al. 2006. J. Bone Miner. Res.21, 934.
Niehrs, C. 2006. Oncogene25, 7469.
Verani, R. et al. 2006. J. Neurochem.101, 242.
Cappuccio, I. et al. 2005. J. Neurosci.25, 2647.
Kemp, C. et al. 2005. Dev. Dyn.233, 1064.
Adamska, M. et al. 2004. Dev. Biol.272, 134.
Bullock, C.M. et al. 2004. Mol. Pharmacol.65, 582.
Mao, B. et al. 2002. Nature417, 664.
Mao, B. et al. 2001. Nature411, 321.
Glinka, A. et al. 1998. Nature391, 357.
Li, J. et al. 2006. Bone36, 754.
Morvan, F. et al. 2006. J. Bone Miner. Res.21, 934.
Niehrs, C. 2006. Oncogene25, 7469.
Verani, R. et al. 2006. J. Neurochem.101, 242.
Cappuccio, I. et al. 2005. J. Neurosci.25, 2647.
Kemp, C. et al. 2005. Dev. Dyn.233, 1064.
Adamska, M. et al. 2004. Dev. Biol.272, 134.
Bullock, C.M. et al. 2004. Mol. Pharmacol.65, 582.
Mao, B. et al. 2002. Nature417, 664.
Mao, B. et al. 2001. Nature411, 321.
Glinka, A. et al. 1998. Nature391, 357.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
HIS TAG is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service