Skip to Content
Merck
All Photos(1)

Key Documents

SRE0066

Sigma-Aldrich

Guanidine hydrochloride solution, 6M

Synonym(s):

Aminoformamidine hydrochloride, GuHCl, Guanidinium chloride, Guanidium chloride

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352209
NACRES:
NA.25

form

liquid

Quality Level

concentration

6 M

application(s)

diagnostic assay manufacturing

storage temp.

room temp

Application

Guanidine hydrochloride is commonly used as a denaturant, because of its ability to break hydrogen bonds between amino acid residues. By breaking these bonds, the 3D conformation of the protein is unfolded and the aqueous solubility of the protein is greatly increased. Once denatured, the protein can be easily reduced, modified, or analyzed, in a variety of procedures. Further processing of the sample can allow the proteins to be analyzed by other common protein analysis methods including mass spectrometry, electrophoresis, and enzymatic digests.
Guanidine hydrochloride has also typically been used for the isolation of RNA, to denature globular proteins, and for protein refolding studies. It can also be used to facilitate the generation of tryptic peptides for analysis of complex protein samples.

Physical form

This product is a ready-to-use 6 M guanidine hydrochloride solution. The 6 M Guanidine hydrochloride solution may be used as a stock solution to prepare solutions of lower concentrations as appropriate.

Note: If precipitation is present in the 6 M buffer, warm the bottle to 37 °C and mix until completely dissolved prior to use.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun-Yi Zhu et al.
Autophagy, 17(10), 3160-3174 (2021-01-07)
We investigated in larval and adult Drosophila models whether loss of the mitochondrial chaperone Hsc70-5 is sufficient to cause pathological alterations commonly observed in Parkinson disease. At affected larval neuromuscular junctions, no effects on terminal size, bouton size or number
Georgios I Laliotis et al.
Nature communications, 12(1), 4624-4624 (2021-08-01)
AKT-phosphorylated IWS1 regulates alternative RNA splicing via a pathway that is active in lung cancer. RNA-seq studies in lung adenocarcinoma cells lacking phosphorylated IWS1, identified a exon 2-deficient U2AF2 splice variant. Here, we show that exon 2 inclusion in the
Kaamini M Dhanabalan et al.
Biomaterials science, 8(15), 4308-4321 (2020-07-01)
Osteoarthritis (OA) is a joint disease characterized by progressive damage of articular cartilage and the adjoining subchondral bone. Chondrocytes, the primary cells of the cartilage, have limited regenerative capacity and when they undergo stress due to trauma or with aging
Yoshinori Aso et al.
eLife, 8 (2019-11-15)
Animals employ diverse learning rules and synaptic plasticity dynamics to record temporal and statistical information about the world. However, the molecular mechanisms underlying this diversity are poorly understood. The anatomically defined compartments of the insect mushroom body function as parallel
Jingwen Yang et al.
Science signaling, 14(665) (2021-01-14)
Cranial neural crest cells (CNCCs) are a population of multipotent stem cells that give rise to craniofacial bone and cartilage during development. Bone morphogenetic protein (BMP) signaling and autophagy have been individually implicated in stem cell homeostasis. Mutations that cause

Articles

A RPLC-MS workflow for mass analysis of reduced monoclonal antibody, trastuzumab, including reduction procedures, calibration, and system suitability tests.

A RPLC-MS workflow for mass analysis of reduced monoclonal antibody, trastuzumab, including reduction procedures, calibration, and system suitability tests.

A RPLC-MS workflow for mass analysis of reduced monoclonal antibody, trastuzumab, including reduction procedures, calibration, and system suitability tests.

A RPLC-MS workflow for mass analysis of reduced monoclonal antibody, trastuzumab, including reduction procedures, calibration, and system suitability tests.

See All

Protocols

SEC-MS protocol for rapid glycoprofiling of monoclonal antibodies with antibody purification and mass spectrometer calibration.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service