Z275751
Nalgene® pipette jars
size E, max. pipette length 600 mm 24 in.
Synonym(s):
pipet jar, pipette storage
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122800
NACRES:
NB.01
Recommended Products
material
high-density polyethylene
size
E
feature
Holds 600 mm pipettes
packaging
pack of 1 ea
manufacturer/tradename
Nalgene® 5242-0050
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Resilient HDPE container for soaking pipettes. Also make excellent general-purpose containers. All sizes have a flanged base for added stability and a dust cover.
CAUTION: Concentrated sulfuric acid will cause pipette jar to fail. Keep the acid concentration at 60% or less.
CAUTION: Concentrated sulfuric acid will cause pipette jar to fail. Keep the acid concentration at 60% or less.
Application
Nalgene® pipette jars have been used for sterilization of glass pipettes.
Features and Benefits
Nalgene® pipette jars have the following benefits:
- The lid enables slow evaporation of the cleaning solution.
- The jar is chemically compatible with extended exposure to various acids, bases, detergents, and alcohols.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service