Skip to Content
Merck
All Photos(3)

Documents

W208100

Sigma-Aldrich

Isoamyl phenylacetate

greener alternative

natural, ≥98%, FG

Synonym(s):

3-methylbutyl 2-phenylacetate, Iso amyl alpha-toluate, Phenyl acetic acid isopentyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H18O2
CAS Number:
Molecular Weight:
206.28
FEMA Number:
2081
EC Number:
Council of Europe no.:
2161
MDL number:
UNSPSC Code:
12164502
Flavis number:
9.789
NACRES:
NA.21

grade

FG
Fragrance grade
Halal
Kosher
natural

Quality Level

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥98%

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.485 (lit.)

bp

268 °C (lit.)

density

0.975 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

greener alternative category

Organoleptic

honey; rose; sweet

SMILES string

CC(C)CCOC(=O)Cc1ccccc1

InChI

1S/C13H18O2/c1-11(2)8-9-15-13(14)10-12-6-4-3-5-7-12/h3-7,11H,8-10H2,1-2H3

InChI key

QWBQBUWZZBUFHN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Isoamyl phenylacetate, a phenethyl alcohol derivative, is commonly used as a flavor additive in tobacco. It is one of the volatile compounds found in apple pomace.
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is Biobased and thus aligns with "Less Hazardous Chemical Syntheses" and "Use of Renewable Feedstocks".

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

217.9 °F

Flash Point(C)

103.3 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of Volatile Compounds in Apple Pomace by Stir Bar Sorptive Extraction and Gas Chromatography-Mass Spectrometry (SBSE-GC-MS).
Madrera RR and Valles BS
Journal of Food Science, 76(9) (2011)
Qualitative and Quantitative Analysis of Flavor Additives on Tobacco Products Using SPME- GC- Mass Spectroscopy
Clark TJ and Bunch JE
Journal of Agricultural and Food Chemistry, 45(3), 844-849 (1997)
T B Adams et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 43(8), 1179-1206 (2005-06-14)
This publication is the ninth in a series of safety evaluations performed by the Expert Panel of the Flavor and Extract Manufacturers Association (FEMA). In 1993, the Panel initiated a comprehensive program to re-evaluate the safety of more than 1700

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service