Merck
  • The FEMA GRAS assessment of phenethyl alcohol, aldehyde, acid, and related acetals and esters used as flavor ingredients.
The FEMA GRAS assessment of phenethyl alcohol, aldehyde, acid, and related acetals and esters used as flavor ingredients.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2005-06-14)
T B Adams, S M Cohen, J Doull, V J Feron, J I Goodman, L J Marnett, I C Munro, P S Portoghese, R L Smith, W J Waddell, B M Wagner
ABSTRACT
This publication is the ninth in a series of safety evaluations performed by the Expert Panel of the Flavor and Extract Manufacturers Association (FEMA). In 1993, the Panel initiated a comprehensive program to re-evaluate the safety of more than 1700 GRAS flavoring substances under conditions of intended use. Elements that are fundamental to the safety evaluation of flavor ingredients include exposure, structural analogy, metabolism, pharmacokinetics and toxicology. Flavor ingredients are evaluated individually and in the context of the available scientific information on the group of structurally related substances. Scientific data relevant to the safety evaluation of the use of phenethyl alcohol, aldehyde, acid, and related acetals and esters as flavoring ingredients is evaluated. The group of phenethylalcohol, aldehyde, acid, and related acetals and esters was reaffirmed as GRAS (GRASr) based, in part, on their self-limiting properties as flavoring substances in food, their rapid absorption, metabolic detoxication, and excretion in humans and other animals, their low level of flavor use, the wide margins of safety between the conservative estimates of intake and the no-observed-adverse effect levels determined from subchronic and chronic studies and the lack of significant genotoxic and mutagenic potential. This evidence of safety is supported by the fact that the intake of phenethyl alcohol, aldehyde, acid, and related acetals and esters as natural components of traditional foods is greater than their intake as intentionally added flavoring substances.
Sigma-Aldrich
Phenethyl alcohol, natural, ≥99%, FCC, FG
Sigma-Aldrich
Phenethyl alcohol, ≥99%, FCC, FG
Supelco
Phenylethyl Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phenethyl propionate, ≥98%, FG
Sigma-Aldrich
2-Phenylethanol, ≥99.0% (GC)
Sigma-Aldrich
Isoamyl phenylacetate, natural, ≥98%, FG