Merck
  • Evaluation of protective efficacy of live attenuated Salmonella enterica serovar Gallinarum vaccine strains against fowl typhoid in chickens.
Evaluation of protective efficacy of live attenuated Salmonella enterica serovar Gallinarum vaccine strains against fowl typhoid in chickens.
Clinical and vaccine immunology : CVI (2014-07-06)
Paweł Laniewski, Arindam Mitra, Kemal Karaca, Ayub Khan, Rajeev Prasad, Roy Curtiss, Kenneth L Roland
RÉSUMÉ
Salmonella enterica serovar Gallinarum is the etiological agent of fowl typhoid, which constitutes a considerable economic problem for poultry growers in developing countries. The vaccination of chickens seems to be the most effective strategy to control the disease in those areas. We constructed S. Gallinarum strains with a deletion of the global regulatory gene fur and evaluated their virulence and protective efficacy in Rhode Island Red chicks and Brown Leghorn layers. The fur deletion mutant was avirulent and, when delivered orally to chicks, elicited excellent protection against lethal S. Gallinarum challenge. It was not as effective when given orally to older birds, although it was highly immunogenic when delivered by intramuscular injection. We also examined the effect of a pmi mutant and a combination of fur deletions with mutations in the pmi and rfaH genes, which affect O-antigen synthesis, and ansB, whose product inhibits host T-cell responses. The S. Gallinarum Δpmi mutant was only partially attenuated, and the ΔansB mutant was fully virulent. The Δfur Δpmi and Δfur ΔansB double mutants were attenuated but not protective when delivered orally to the chicks. However, a Δpmi Δfur strain was highly immunogenic when administered intramuscularly. All together, our results show that the fur gene is essential for the virulence of S. Gallinarum, and the fur mutant is effective as a live recombinant vaccine against fowl typhoid.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Saccharose, for molecular biology, ≥99.5% (GC)
Référence
Conditionnement
Disponibilité
Prix
Quantité
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Référence
Conditionnement
Disponibilité
Prix
Quantité
Sigma-Aldrich
Chloramphénicol, ≥98% (HPLC)
Référence
Conditionnement
Disponibilité
Prix
Quantité
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioXtra
Référence
Conditionnement
Disponibilité
Prix
Quantité
Sigma-Aldrich
Saccharose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Référence
Conditionnement
Disponibilité
Prix
Quantité
USP
Saccharose, United States Pharmacopeia (USP) Reference Standard
Référence
Conditionnement
Disponibilité
Prix
Quantité
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Référence
Conditionnement
Disponibilité
Prix
Quantité
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Référence
Conditionnement
Disponibilité
Prix
Quantité
Sigma-Aldrich
Saccharose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Référence
Conditionnement
Disponibilité
Prix
Quantité
Sigma-Aldrich
D-(+)-Mannose, powder, BioReagent, suitable for cell culture
Référence
Conditionnement
Disponibilité
Prix
Quantité
Sigma-Aldrich
Chloramphénicol, BioReagent, suitable for plant cell culture
Référence
Conditionnement
Disponibilité
Prix
Quantité
Sigma-Aldrich
D-(+)-Mannose, synthetic, ≥99% (GC)
Référence
Conditionnement
Disponibilité
Prix
Quantité
Sigma-Aldrich
Saccharose, Grade I, ≥99% (GC), suitable for plant cell culture
Référence
Conditionnement
Disponibilité
Prix
Quantité
Sigma-Aldrich
Tetrazolium Blue Chloride, indicator for germination, suitable for microbiology, ≥90% (T)
Référence
Conditionnement
Disponibilité
Prix
Quantité
Sigma-Aldrich
D-(−)-Arabinose, ≥98% (GC)
Référence
Conditionnement
Disponibilité
Prix
Quantité
Sigma-Aldrich
Saccharose, meets USP testing specifications
Référence
Conditionnement
Disponibilité
Prix
Quantité
Sigma-Aldrich
D-(+)-Mannose, ≥99% (GC), wood
Référence
Conditionnement
Disponibilité
Prix
Quantité
Sigma-Aldrich
Chloramphénicol, meets USP testing specifications
Référence
Conditionnement
Disponibilité
Prix
Quantité
Sigma-Aldrich
D-(+)-Mannose, BioUltra, ≥99.5% (sum of enantiomers, HPLC)
Référence
Conditionnement
Disponibilité
Prix
Quantité
Supelco
Saccharose, Pharmaceutical Secondary Standard; Certified Reference Material
Référence
Conditionnement
Disponibilité
Prix
Quantité
Sigma-Aldrich
Saccharose, ACS reagent
Référence
Conditionnement
Disponibilité
Prix
Quantité
Millipore
Saccharose, suitable for microbiology, ACS reagent, ≥99.0%
Référence
Conditionnement
Disponibilité
Prix
Quantité
Sigma-Aldrich
Saccharose, puriss., meets analytical specification of Ph. Eur., BP, NF
Référence
Conditionnement
Disponibilité
Prix
Quantité
Sigma-Aldrich
Tetrazolium Blue Chloride, used in colorimetric determination of reducing compounds
Référence
Conditionnement
Disponibilité
Prix
Quantité
Supelco
Chloramphénicol, VETRANAL®, analytical standard
Référence
Conditionnement
Disponibilité
Prix
Quantité
Millipore
D-(+)-Mannose, ≥99%, suitable for microbiology
Référence
Conditionnement
Disponibilité
Prix
Quantité
Saccharose, European Pharmacopoeia (EP) Reference Standard
Référence
Conditionnement
Disponibilité
Prix
Quantité
Supelco
Saccharose, analytical standard, for enzymatic assay kit SCA20
Référence
Conditionnement
Disponibilité
Prix
Quantité
Millipore
D-(−)-Arabinose, suitable for microbiology, ≥99.0%
Référence
Conditionnement
Disponibilité
Prix
Quantité
Chloramphénicol, European Pharmacopoeia (EP) Reference Standard
Référence
Conditionnement
Disponibilité
Prix
Quantité