Merck
Various forms of rabbit plasma alpha-1-antiproteinase.
Biochemistry and molecular biology international (1998-10-24)
A Saito, H Sinohara
RÉSUMÉ
Amino acid sequencing of the ficin-derived C-terminal fragments of alpha-1-antiproteinase (also called alpha-1-antitrypsin or alpha-1-proteinase inhibitor) prepared from rabbit plasma revealed the presence of the E isoform, which had been confirmed in the cDNA library in addition to the F and S-1 forms. The S-2 form was identified in inflamed rabbit plasma. The ficin digest of human plasma alpha-1-antitrypsin resulted in a major fragment of the M type. Multiple forms of alpha-1-antiproteinase in the rabbit plasma implicate the unknown functions other than the inhibition of neutrophil elastase.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Ficin from fig tree latex, lyophilized powder
Sigma-Aldrich
Ficin from fig tree latex, powder, ≥0.1 unit/mg solid
Sigma-Aldrich
Ficin from fig tree latex, saline suspension, ≥1.0 units/mg protein (biuret)