Principaux documents
52818-U
Supelco
HybridSPE®-PLus 96-Well Plate Essentials Kit
96-Well Essentials Kit, pk of 1
About This Item
Code UNSPSC :
41115712
Nomenclature NACRES :
NB.21
Conditionnement
pk of 1
Technique(s)
solid phase extraction (SPE): suitable
Description générale
HybridSPE-PLus 96-well Plate Essentials Kit includes one each of the following:
  • 96-well HybridSPE-PLus plate (575659-U)
  • 96 round well collection plate, 2 mL, polypropylene
  • 96 round well pierceable cap mat
  • plate sealing film sheet
Informations légales
HybridSPE is a registered trademark of Merck KGaA, Darmstadt, Germany
Composants de kit également disponibles séparément
Réf. du produit
Description
FDS
  • 575659-UHybridSPE®-PLus 96-Well Plate, volume 2 mL, pk of 1 eaFDS
  • 96 Round Well Pierceable Cap Mats, for use with HybridSPE-PLus 96-well plates, pk of 50 ×
  • Z721581EZ-Pierce films, Excel Scientific EZP-100, non-sterileFDS
  • Z717266Nunc® 96 DeepWell plate, non-treated, maximum volume 2 mL, non-sterile, round bottom natural polypropylene wellsFDS
Articles
This article highlights the impact that sample matrix effects can have on LC/MS response and discusses two novel approaches to reduce it.
Protocoles
Immunoassay-based methods for digitoxin or digoxin exist, but are time consuming, labor intensive, and not selective due to similarities in their chemical structures.
The conversion of clinical methods to LC/MS/MS offers advantages; however, is accompanied by a few limitations, notably interference effects from the endogenous sample matrix.