Principaux documents
27147
Supelco
Flacons (vials), bouchon à vis à ouverture centrale phénolique, pré-assemblés
volume 7 mL, clear glass vial, O.D. × H 17 mm × 60 mm, PTFE/red rubber septum
About This Item
Code UNSPSC :
41122001
Matériaux
PTFE/red rubber septum
black phenolic cap (open-top)
clear glass vial
Caractéristiques
closure type black phenolic hole cap
closure type screw top vial
Conditionnement
pkg of 100 ea
D.E. × H
17 mm × 60 mm
Volume
7 mL
Raccords
thread for 15-425
Application
It may be used in headspace solid-phase microextraction (SPME) followed by GC-FID for simultaneous analysis of furan, 2-methylfuran and 2-pentylfuran from juice samples.
An efficient method for the simultaneous determination of furan, 2-methylfuran and 2-pentylfuran in fruit juices by headspace solid phase microextraction and gas chromatography?flame ionisation detector.
Hu, Gaofei, et al.
Food Chemistry, 192, 9-14 (2016)