Principaux documents
23632
Supelco
Connecteur GlasSeal en "Y", silice fondue
pkg of 3 ea
3 EA
341,00 €
341,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Devis pour commande en gros
Changer de vue
3 EA
341,00 €
About This Item
Code UNSPSC :
41115700
341,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Devis pour commande en gros
Conditionnement
pkg of 3 ea
Technique(s)
gas chromatography (GC): suitable
Description générale
Quickly connect three pieces of fused silica tubing of the same or different diameters without the need for tools. The leak-free connection is useful to split a sample to two columns, or to split a column effluent to two detectors. Silanized for an inert inside surface. For use with 0.10 - 0.53 mm I.D. fused silica tubing.
Application
"Y" GlasSeal Connector maybe used in capillary gas chromatographic-electroantennographic detection (GC-EAD) for chirality analysis of essential amino acid methyl esters.[1]
Autres remarques
We offer a variety of chromatography accessories including analytical syringes
Essential amino acid methyl esters: major sex pheromone components of the cranberry white grub, Phyllophaga anxia (Coleoptera: Scarabaeidae).
Zhang A
Journal of Chemical Ecology, 23 (1), 231-245 (1997)