Documents
21019-U
Supelco
LPDNPH Lapel Clips
pkg of 6 ea
About This Item
Code UNSPSC :
12190000
Nomenclature NACRES :
NB.24
Conditionnement
pkg of 6 ea
Technique(s)
active air sampling: suitable
Application(s)
air monitoring
environmental
industrial hygiene
Compatibilité
LpDNPH cartridges