Principaux documents
2051701
Supelco
Direct
pkg of 1 ea
About This Item
Code UNSPSC :
41121800
Conditionnement
pkg of 1 ea
L × D.E. × D.I.
78.5 mm × 6.3 mm × 1.5 mm
Description générale
Deactivated glass liner for capillary injection port
Inlet liner is an important part within the injection port. The prime functions of the inlet liner are to limit sample degradation and improve vaporisation along with correctly positioning the syringe needle. A wrong choice of liner can have prominent effect on the chromatogram. The basic problems related with incorrect choice of liner include the higher potential of peak tailing and mass discrimination. The inlet liner volume is recommended to be larger than the amount of sample injected by syringe so as to avoid any reaction with the sample. Injector port liner is suitable to make connection between sample introduction and GC column. Direct type has narrow bore which sustains a high linear velocity through the injection port to diminish band broadening and transferring all the samples to column.
Application
Inlet liner, direct type may be used for solid phase microextraction (SPME) sample injection.
Novel aggregation pheromone,(1S, 4R)-p-menth-2-en-1-ol, of the ambrosia beetle, Platypus quercivorus (Coleoptera: Platypodidae)
Tokoro, Masahiko, et al.
Bulletin of the Forestry and Forest Products Research Institute, 6, 49-57 (2007)
Michelle Groves Carlin, John Richard Dean
Forensic Applications of Gas Chromatography, 9-9 (2013)