Documents
Z743487
Aluminum heating/cooling block
for 384-well plates, 384-wells in centre, 10 on sides (4 x 1.5 mL & 6 x 0.5 mL)
Synonyme(s) :
dry block, plate cooling block, plate heating block, tube cooling block, tube heating block
About This Item
Code UNSPSC :
41102404
Nomenclature NACRES :
NB.77
Caractéristiques
for 384-well plates, 384-wells in centre, 10 on sides (4 x 1.5 mL & 6 x 0.5 mL)
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Diversified Biotech CHAM-3840
Description générale
Each Cooling Chamber is a solid aluminum block that surrounds inserted tubes to guarantee sustained hot or cold temperatures.