Documents
Z740348
Eppendorf® Reference® 2 Fixed Volume Pipettor
volume 250 μL, blue
About This Item
Code UNSPSC :
41121500
Nomenclature NACRES :
NB.01
Matériaux
blue
Caractéristiques
electronic: no
single-channel
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Eppendorf® 4925000138
Volume
250 μL
Description générale
The Reference 2 is the successor to the highly successful Reference pipette, and enhances the range′s already high standards with a new design, reduced weight and operating forces and multi-channel versions. As the premium product in Eppendorf′s pipette portfolio, the Reference 2 provides the most accurate results possible alongside robust, reliable handling and optimum user safety.
These levels of security make the Reference 2 ideal for use with precious liquids, or in any application requiring extremely high levels of accuracy. When there can be no compromise on quality and reproducability, Eppendorf′s new Reference 2 is an ideal instrument for these liquid handling applications.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG
Reference is a registered trademark of Eppendorf AG