Principaux documents
Z692603
microDOC Compact gel documentation system
AC/DC input 110 V AC
1 EA
4 230,00 €
4 230,00 €
Date d'expédition estimée le29 avril 2025Détails
Changer de vue
1 EA
4 230,00 €
About This Item
Code UNSPSC :
41105300
Nomenclature NACRES :
NB.22
4 230,00 €
Date d'expédition estimée le29 avril 2025Détails
Fabricant/nom de marque
Cleaver CSL-MICRODOC$
Description générale
The microDOC Compact Gel Documentation System gives you all of the advantages of the Basic system with the advantage of a large 8 inch TFT colour monitor allowing clear and simple image visualization. Can be used with most 12 x 12 cm tranilluminators.
Available with optional TotalLab 1D software. Main software functions include: lane creation, background subtraction, band detection, profile deconvolution, and molecular size, pI and quantity calibration.
System comprises of:
Canon G12 digital
2MB storage card
Ethidium bromide & close up +3 filters
Lift-off hood with easy access door and built-in inner lights
8 in. TFT color monitor
Informations légales
TotalLab is a trademark of Nonlinear Dynamics Ltd.