Principaux documents
Z692557
microDOC Basic gel documentation system
AC/DC input 110 V AC
1 EA
2 600,00 €
2 600,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
2 600,00 €
About This Item
Code UNSPSC :
41105300
2 600,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Fabricant/nom de marque
Cleaver CSL-MDOCBASIC$
Description générale
System contains: Canon G12 digital camera, basic lift-off dark room hood.
This is the perfect low cost option to upgrade to digital documentation of your gels. Can be used with most 21 x 21 cm tranilluminators.
Main software functions include: lane creation, background subtraction, band detection, profile deconvolution, and molecular size, pI and quantity calibration.
System comprises of:
  • Canon G15 digital camera (12.1 MP)
  • 2GB storage card
  • Ethidium bromide & close up +3 filters
  • Lift-off hood
  • Available with optional TotalLab 1D software
Informations légales
TotalLab is a trademark of Nonlinear Dynamics Ltd.