Z683191
Caisse d'expédition pour petits rongeurs
About This Item
Produits recommandés
L × l × P
17.75 in. × 9 in. × 6 in., 159 total square inches. Configurable up to 2 compartments.
Application
- Spunbond Polypropylene - 170 cfm/ft
Autoclave Cycles:
- 2 cycles maximum recommended up to 126 ºC, 20 minutes maximum
- Meets IATA (International Air Transport Association) specifications
- All-plastic construction: box, lid, dividers and filter medium
- Superior airflow via patented engineered openings
- Easily stackable for material handling and transportation, with integral spacers to ensure proper air circulation
- Clear plastic windows enables at-a-glance inspection of lab animals
- Positive open/close lid facilitates animal insertion/removal
- Interior rim provides a barrier against gang and go incidents
- Manufactured in the USA
- 9Chloroform
- Clorosulfonic Acid
- Chlorine Gas
- Tétrachlorure de carbone
- Ixooctane
- Xylène
- Acide nitrique
- Éther de pétrole
- Chlorure d'amyle
- Acétate de butyle
- Benzène
- Eau de brome
- Most Petroleum fuels will attack
Caractéristiques et avantages
- Sigma-Aldrich offers plastic containers for shipping your research models more safely. These containers are sturdier than cardboard boxes and have excellent protection against contaminants, breakage, and leakage en route to your lab
- The patented design also ensures unsurpassed convenience and ease of handling at every stage
Composants de kit seuls
- Container 1 ea
- Lid 1 ea
- Large Closure Label 1 ea
- Small Closure Label 2 ea
- Live Animal Label 2 ea