Z380377
Nalgene® Griffin beakers
capacity 2,000 mL, translucent polypropylene
Synonyme(s) :
PP beaker, nalgene PP beaker
About This Item
Produits recommandés
Matériaux
translucent polypropylene
Caractéristiques
graduations
Conditionnement
pkg of 1
Fabricant/nom de marque
Nalgene 1201-2000
Disponibilité
not available in EU
Capacité
2,000 mL
Catégories apparentées
Description générale
Low form. With silk-screened graduations, Nalgene catalog number, size and resin codes, max. use temp., and "no flame" symbol. Meet the ISO/DIS 7056 International Standards for Laboratory Plasticware Accuracy. Chemically resistant. Autoclavable. Not for use on a hot plate.
Autres remarques
Browse all plastic beakers
Informations légales
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries