Principaux documents
BR55646
BRAND® conical joint clip
polyoxymethylene, for NS 29/32
Synonyme(s) :
Conical joint stopper
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NB.51
Matériaux
polyoxymethylene
red
Description
Operating temperature up to 150 °C
Conditionnement
pack of 1 ea
Fabricant/nom de marque
BRAND 55646
Description générale
Will not scratch glass surfaces. Excellent resilience and stability. Outstanding chemical resistance.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG