Principaux documents
Z332259
Bottle adapters for BRAND® Dispensers and Burettes
GL 32 thread, bottle (NS 19/26), polypropylene
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.01
Matériaux
polypropylene
Fabricant/nom de marque
BRAND 704419
Raccords
bottle (NS 19/26)
GL 32 thread
Catégories apparentées
Description générale
For all Dispensette® models as well as seripettor® and QuikSip
Autres remarques
BR704419 (India)
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
QuikSip is a trademark of BRAND GMBH + CO KG
seripettor is a registered trademark of BRAND GMBH + CO KG