Z329991
BRAND® narrow-mouth bottle, LDPE
capacity 1000 mL
Synonyme(s) :
ldpe bottle, polyethylene bottles, polyethylene containers
125,00 €
About This Item
125,00 €
Produits recommandés
Matériaux
low-density polyethylene bottle
narrow-mouth bottle
round bottle
translucent
Joint
Joints threaded neck (GL28)
Description
dropper nozzle caps available
Caractéristiques
with cap
Conditionnement
pkg of 25
Fabricant/nom de marque
BRAND 129462
Capacité
1000 mL
Diam. × H
94 mm × 212 mm