Z326879
BRAND® BLAUBRAND® volumetric flask, glass stopper, clear glass
volume 25 mL, accuracy: 0.04 mL, neck joint: ST/NS 10/19, glass stopper
44,50 €
About This Item
44,50 €
Produits recommandés
Matériaux
borosilicate glass
glass stopper
Joint
joint: ST/NS neck10/19
Stérilité
non-sterile
Caractéristiques
class A DE-M, DIN EN ISO 1042. Calibrated to contain (TC, In).
closure type Glass stopper
Conditionnement
pack of 2
Fabricant/nom de marque
BRAND
Plage de mesure du volume
25 mL, accuracy: 0.04 mL
Capacité
25 mL
Description générale
All BLAUBRAND® volumetric flasks are supplied with a batch number and an accompanying batch certificate in the original packaging.