Principaux documents
Z324884
Sigma-Aldrich
BRAND® SILBERBRAND ETERNA volumetric pipettes, calibrated to deliver
capacity 50 mL
About This Item
Code UNSPSC :
41121500
Nomenclature NACRES :
NB.01
Matériaux
AR-Glas®
Agence
according to DIN 12 690
Caractéristiques
class B (callibrated to deliver)
class B
one-mark
Conditionnement
pack of 6 ea
Fabricant/nom de marque
BRAND 29518
Plage de mesure du volume
50 mL, accuracy: 0.07 mL
Capacité
50 mL
Description générale
The BRAND® SILBERBRAND ETERNA volumetric pipettes are expertly crafted from AR-Glas® and conform to DIN EN ISO 648 standards, ensuring high precision and reliability. Calibrated to deliver (TD, Ex), these pipettes feature a waiting time of 5 seconds for optimal accuracy in liquid transfer. Each BLAUBRAND® bulb pipette is supplied with a batch number and includes one batch certificate per packing unit from the manufacturer, guaranteeing traceability and quality. This combination of quality materials and thorough documentation makes these pipettes an excellent choice for demanding laboratory environments.
Autres remarques
BR29518 (India)
Informations légales
AR-Glas is a registered trademark of Schott AG
BRAND is a registered trademark of BRAND GMBH + CO KG
Protocoles
This page contains detailed information on working with and handling pipettes and pipetting aids.