Principaux documents
TMO4114-0038
Nalgene® vented sterile caps for Erlenmeyer flasks
high-density polyethylene, 125/250 mL flasks, pore size 0.2 μm, case of 12 ea
Synonyme(s) :
vented closure, vented single use closure
1 EACH
109,00 €
109,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EACH
109,00 €
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.13
109,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
high-density polyethylene
Stérilité
sterile
Caractéristiques
vented
Conditionnement
case of 12 ea
Fabricant/nom de marque
Nalgene 4114-0038
Dimension de pores
0.2 μm pore size
Description générale
Ensure a sterile gas exchange for shaker and suspension cell culture by using Thermo Scientific Nalgene Vented HDPE Closures for Sterile Single Use Erlenmeyer Flasks.
Hold and sterilize refuse, clothing, equipment, labware and contaminated material with Thermo Scientific Nalgene Large Polypropylene Waste Containers with Cover. These containers feature a secure, friction-fit closure and molded-in handgrips. They meet OSHA Standard 29 CFR Part 1910.1030 for use as protection against bloodborne pathogens.
Informations légales
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries