Principaux documents
TMO382099-0250
Nalgene® Certified Low Particulate narrow-mouth bottle
natural high-density polyethylene, volume 250 mL, case of 72 ea
Synonyme(s) :
HDPE bottle, certified nalgene bottle
72 EACH
362,00 €
362,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
72 EACH
362,00 €
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
362,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
natural high-density polyethylene
Conditionnement
case of 72 ea
Fabricant/nom de marque
Nalgene 382099-0250
Capacité
250 mL
Volume
250 mL
Description générale
Effectively store high-purity chemicals with Thermo Scientific Nalgene Certified Low Particulate Narrow-Mouth HDPE Bottles with Closure, which are extremely low in trace metal content. Controlled environment manufacturing minimizes particulate contamination.
Informations légales
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries