Principaux documents
TMO312089-0001
Nalgene® narrow-mouth bottle
natural high-density polyethylene, size 30 mL, case of 1000 ea
Synonyme(s) :
Nalgene bottle, plastic bottle
1000 EACH
772,00 €
772,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1000 EACH
772,00 €
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
772,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
natural high-density polyethylene
Conditionnement
case of 1000 ea
Fabricant/nom de marque
Nalgene 312089-0001
Taille
30 mL
Description générale
Store and ship light-sensitive reagents with Thermo Scientific Nalgene Narrow-Mouth Opaque Amber HDPE Packaging Bottles with Closure. Ideal for diagnostics, biologicals, specialty chemicals, reagents, adhesives and veterinary pharmaceuticals, these high density polyethylene bottles are injection blow-molded for uniform wall thickness and strength. They meet USP light transmission standards and are bulk packed for packaging applications.
Informations légales
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries