This product has a working volume of 1.5 mL per well and a culture area of 3.14 cm2. The membrane has a pore density of <0.85 x 108 pores/cm2.
TMO140640
Nunc® cell culture inserts in 6-well Nunclon(TM) Delta surface plate
cell culture treated, polycarbonate, sterile, pore size 0.4 μm, case of 24 ea (supplied in 4 x 6 well plates)
Synonyme(s) :
multidish membrane insert, treated membrane insert
278,00 €
About This Item
278,00 €
Produits recommandés
Matériaux
polycarbonate
Stérilité
sterile
Conditionnement
case of 24 ea (supplied in 4 x 6 well plates)
Fabricant/nom de marque
Nunc 140640
Dimension de pores
0.4 μm pore size
Type de liaison
cell culture treated
-
ich habe 6-wells plate for transwell assay bestellt. aber ich weiss nicht wie viel medium sollte ich in einen Well benutzen, um alle Zellen zu erreichen. Gibt es einen Protocol?