Principaux documents
PCRTUBES-EP
Eppendorf® PCR tubes
colorless, capacity 0.2 mL, pack of 1000
About This Item
Code UNSPSC :
41121703
Le produit PCRTUBES-EP n'est pas actuellement commercialisé dans votre pays Contacter notre Service technique
Matériaux
colorless
conical bottom polypropylene
Stérilité
non-sterile
Caractéristiques
RNase and DNase free
cap (hinged lid clip)
Conditionnement
pack of 1000
Fabricant/nom de marque
Eppendorf® 951010006 (US)
Technique(s)
PCR: suitable
Capacité
0.2 mL
Description générale
Eppendorf® PCR tubes adhere to rigorous quality standards set by Eppendorf. Their simple design guarantees an airtight seal, preventing contamination and evaporation, while allowing for effortless one-handed operation. The tubes′ thin polypropylene walls enable efficient and uniform heat transfer, leading to fast andhigh-quality PCR outcomes. The PCR clean purity level is maintained through adual-layer packaging system comprising resealable bags inside the outer carton. The consistent wall thickness and smooth surface of the Eppendorf® PCR tubes facilitate efficient sample heating, accelerating the PCR process.
Caractéristiques et avantages
  • The hinged lid features a contamination shield.
  • The lid has a special hinge that ensures a defined position.
  • The tube base maintains high transparency.
  • The lid is etched for convenient labeling.
  • Provides a tight seal to prevent leakage.
  • Easy to open.
  • Compatible with all thermal cyclers using the 0.2 mL block format.
  • Also offered in an 8-tube strip format.
  • Batch tested and certified to be free of DNA, DNase, RNase, and PCR inhibitors (PCR clean).
Informations légales
Eppendorf is a registered trademark of Eppendorf AG