Principaux documents
PCG3011
Sigma-Aldrich
TruPAGE Transfer Buffer
20 ×
About This Item
Code UNSPSC :
12352201
Nomenclature NACRES :
NB.22
Concentration
20 ×
Compatibilité
for use with Sigma-Aldrich TruPAGE Precast Gels
Température de stockage
2-8°C
Application
TruPAGE Transfer Buffer is specially formulated for protein transfer to PVDF or Nitrocellulose membranes from TruPAGE Precast Gels.[1][2]
Caractéristiques et avantages
TruPAGE gel cassettes have a hassle-free design:
  • Extra tall wells to prevent lane-to-lane overflow.
  • No comb to remove, preventing torn or bent wells. Instead, the cassettes lock well fingers in place.
  • No tape to remove
TruPAGE Transfer Buffer is specially formulated for protein transfer to PVDF or Nitrocellulose membranes from TruPAGE Precast Gels.
Autres remarques
Diluted as 20X solution for use with the XCell and BioRad Blotting apparatuses.
Informations légales
TruPAGE is a trademark of Sigma-Aldrich Co. LLC
Code de la classe de stockage
12 - Non Combustible Liquids
Classe de danger pour l'eau (WGK)
WGK 1
Point d'éclair (°F)
Not applicable
Point d'éclair (°C)
Not applicable
Registered report: Oncometabolite 2-hydroxyglutarate is a competitive inhibitor of a-ketoglutarate-dependent dioxygenases.
Evans B and Erin G
eLife, 4, e07420-e07420 (2015)
Guoxia Zhu et al.
Neural plasticity, 2020, 8823785-8823785 (2020-10-22)
Sound conditioning (SC) is defined as "toughening" to lower levels of sound over time, which reduces a subsequent noise-induced threshold shift. Although the protective effect of SC in mammals is generally understood, the exact mechanisms involved have not yet been
Registered report: Transcriptional amplification in tumor cells with elevated c-Myc.
Blum D, et al.
eLife, 4, e04024-e04024 (2015)
Brad Evans et al.
eLife, 4, e07420-e07420 (2015-08-01)
The Reproducibility Project: Cancer Biology seeks to address growing concerns about reproducibility in scientific research by conducting replications of selected experiments from a number of high-profile papers in the field of cancer biology. The papers, which were published between 2010
Eleanor Wendy Trotter et al.
Open biology, 10(10), 200200-200200 (2020-10-15)
Each approach used to synchronize cell cycle progression of human cell lines presents a unique set of challenges. Induction synchrony with agents that transiently block progression through key cell cycle stages are popular, but change stoichiometries of cell cycle regulators