HS81121C
Inoculating Needle/Loops
loop volume 10 μL, flexible
, sterile, blue
Synonyme(s) :
Inoculation Tools, Sterile Inoculating Loops
About This Item
Produits recommandés
Matériaux
blue
Stérilité
sterile
Conditionnement
pack of 1000 ea
Fabricant/nom de marque
Heathrow Scientific HS81121C
Volume de la boucle
10 μL
Description générale
The Inoculating Needle/Loops combination provides choice to spread or pick colonies and cultures. Transfer bacterial cultures without risk of contamination.
Caractéristiques et avantages
- Use loop end to uniformly streak a gel surface
- Use needle end for removal of colonies
- Constructed of flexible polypropylene
- Sterile, 20 per zip bag with lot-to-lot traceability
- Disposable