Principaux documents
HS23471B
ClearlySafe Glove Box Holder
Fits 2 glove boxes, clear acrylic
Synonyme(s) :
glove dispenser
1 EACH
86,70 €
86,70 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EACH
86,70 €
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.51
86,70 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
clear acrylic
Caractéristiques
Holds 2 boxes
Fabricant/nom de marque
Heathrow Scientific HS23471B
Larg. × H × P
295 × 260 × 110 mm
Caractéristiques et avantages
  • All holders fit most of the box brands
  • Mounting screws are provided
  • A polyester foam insert is included in the clear acrylic holders to firmly hold a variety of glove box sizes-snug fit facilitates quick removal of gloves.
  • all holders fit most major brands of boxes
  • mounting screws included
  • includes a polyester foam insert to securely hold a range of glove sizes- snug fit ensures easy removal of gloves
Autres remarques
Gloves to be organized and kept ready for immediate use. Free up benchtop space by mounting holders on the wall for convenient glove dispensing.