Principaux documents
HS120770
Coated Wire Tube Rack
to hold, 48 x 25-30 mm tubes, blue
Synonyme(s) :
Tube holder, bottle rack
1 EACH
51,60 €
51,60 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EACH
51,60 €
About This Item
Code UNSPSC :
41122804
Nomenclature NACRES :
NB.12
51,60 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
blue
high-density polyethylene coating
Caractéristiques
to hold 48 x 25-30 mm tubes
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Heathrow Scientific 120770
L × l × H
10.9 in. (27.7 cm) × 8.3 in. (20.9 cm) × 3.9 in. (9.8 cm)
Description générale
The HDPE Coated Wire Rack is durable for transport or sample handling. It is chemically-resistant and suitable for conical, round or flat-bottom, or plastic tubes and bottles.
Caractéristiques et avantages
  • Chemically-resistant HDPE Coated Wire Racks
  • Racks can be used between temperatures of -40°C and 121°C
  • Holds all brands and types of conical, round or flat bottom, glass or plastic, tubes and/or bottles
  • Square wells and three deck levels help keep tubes secure
  • AutoclavableMesh Bottom Support