GEUS77705
illustra™ ExoProStar 1-Step
500 reactions
Synonyme(s) :
illustra ExoProStar
438,00 €
About This Item
438,00 €
Description générale
illustra ExoProStar is our new name for this PCR clean-up technology. No change in the formulation, so you get the same great product, and same great benefits!
The ExoProStar 1-Step PCR and Sequence Reaction Clean-Up reagent provides a simple, single-step method for removing unincorporated primers and nucleotides prior to downstream analysis. illustra Exonuclease I and Alkaline Phosphatase enzymes are combined in a single, stable mix requiring just one pipetting step to set up the clean-up reaction.
illustra ExoProStar 1-Step is based on GE′s founding patents for the clean-up of PCR reactions using Exonuclease 1 and Alkaline Phosphatase. This 1-step version is not available in USA, Canada and Japan. Instead, we offer a version with the enzymes in two separate tubes, the ExoProStar.
Caractéristiques et avantages
- One tube with one mix of enzymes delivering PCR and sequence reaction clean-up
- Just one pipetting step to set up your reaction
- Fast 30-minute, hands-free protocol
- 100% recovery of PCR product
- Scalable for different reaction sizes
- Automation friendly