Principaux documents
GBL705101
Grace Bio-Labs ONCYTE® SuperNOVA nitrocellulose film slides
1-15 mm x 69 mm NC pad per slide, plain glass
Synonyme(s) :
nitrocellulose slides, protein microarrays
About This Item
Code UNSPSC :
41122600
Matériaux
nitrocellulose (on glass)
Conditionnement
pkg of 20 ea
Fabricant/nom de marque
Grace Bio-Labs 705101
Taille externe
25 mm × 75 mm
Type de liaison
high binding surface
Description générale
Fluorescence detection is a preferred method for many quantitative protein microarray assaysin research and diagnostic applications due to its excellent sensitivity and potential for multiplexing. Use of multiplexed fluorescence-based assays on porous nitrocellulose though has been hampered by high backgrounds at shorter excitation wave lengths (such as the 532nm fluorescence channel). These ONCYTE™ SuperNOVA™ microporous nitrocellulose film slides increase protein microarray assay sensitivity and dynamic range using fluorescent end points at multiple wave lengths. Use of SuperNOVA™ slides can lower the limit of detection with fluorescent end points by 3 – 4 orders of magnitude over competing nitrocellulose film slides.
Informations légales
NOVA is a trademark of Grace Bio-Labs, Inc.
Oncyte is a registered trademark of Grace Bio-Labs, Inc.
SuperNOVA is a trademark of Grace Bio-Labs, Inc.