Principaux documents
EPAG7-TT16
Test Tube Rack, for Eppendorf® New Brunswick Innova® 3100 Water Bath Shakers
holds 36 x 16 mm tubes
1 EA
405,00 €
405,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
405,00 €
About This Item
Code UNSPSC :
41122804
Nomenclature NACRES :
NB.73
405,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
stainless steel
Caractéristiques
holds 36 x 16 mm tubes
Fabricant/nom de marque
Eppendorf® AG7-TT16 (North America)
Description générale
Test Tube Racks have an adjustable angle and tube height ­support. Made from stainless-steel with inserts to help retain tubes and ­prevent them from spinning in the rack. Finger-tight knobs let you quickly adjust angle, or release the rack from the base plate, for easy transport to the bench.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG
Innova is a registered trademark of Eppendorf, Inc. 
New Brunswick is a trademark of Eppendorf AG