Principaux documents
EP4960800147
Eppendorf® Bottle Thread Adapter
from GL 45 to S 40 (buttress thread), polypropylene
1 EA
15,30 €
15,30 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
15,30 €
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.17
15,30 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
polypropylene
Caractéristiques
from GL 45 to S 40 (buttress thread)
Fabricant/nom de marque
Eppendorf® 4960800147
Catégories apparentées
Description générale
Eppendorf dispensers are designed for bottle threads of 45 mm. For other bottle threads a thread adapter can be used. The diameter of the thread adapter is imprinted on the adapter. If the bottle thread is smaller than 32 mm, a second adapter must be used. If you need a thread adapter with a higer resistance to chemicals, use a PTFE/ETFE adapter.
Application
Connection of Eppendorf dispensers to bottles with neck threads different to 45 mm.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG