Principaux documents
EP0030132530
Eppendorf® Twin tec real-time PCR plates
blue plate, conical bottom white wells, semi-skirted, pkg of 25 ea
25 EA
196,00 €
196,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
25 EA
196,00 €
About This Item
Code UNSPSC :
41106300
Nomenclature NACRES :
NB.15
196,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
blue plate
conical bottom white wells
Caractéristiques
skirt (Semi skirted)
Conditionnement
pkg of 25 ea
Fabricant/nom de marque
Eppendorf® 0030132530
Technique(s)
PCR: suitable
Taille
96 well
Volume de travail
250 μL
Description générale
The limiting factor in low-volume real-time PCR (qPCR) is often the remaining fluorescence intensity. Eppendorf twin.tec real-time PCR plates are specially manufactured for qPCR, as they offer all the advantages of regular twin.tec plates, but with the added advantage of white wells. These reflect fluorescence much better than clear or frosted wells, so lower levels of fluorescence are still detectable with the same instrument – just by using the right consumables. The qPCR plates′ white wells also significantly reduce interfering background fluorescence and lead to increased consistency between replicates, and giving highly reproducible results. The wells are made from polypropylene – which guarantees fast heat transfer for fast assays – and the frame is made of rigid polycarbonate, to ensure easy and reliable handling – both in manual and automated assays.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG