Principaux documents
EP0030131517
Eppendorf® plate lid
PCR clean, pkg of 80 ea
80 EA
140,00 €
140,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
80 EA
140,00 €
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.22
140,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Caractéristiques
PCR clean
Conditionnement
pkg of 80 ea
Fabricant/nom de marque
Eppendorf® 003013517
Caractéristiques et avantages
  • Sealing Mats are reusables and can be autoclave. Sealing mats are suitable for short-term storage
    • Heat Sealing Film and Foil for continuous locking give the best evaporation protection
    • Foil products are made of aluminum, can be pierced, and protect light-sensitive samples
    • Film products are transparent and provide protection against unwanted punctures
  • Eppendorf Plate Lid: stable, flexible protection of samples during short-term storage
    • Storage Film and Foil glued seals for simple and dependable sealing during sample storage
Informations légales
Eppendorf is a registered trademark of Eppendorf AG