Principaux documents
EP0030129857
Eppendorf® BioBased twin.tec 96 Well PCR Plate
greener alternative
skirt, (with OptiTrack high contrast labelling)
25 EACH
165,00 €
165,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
25 EACH
165,00 €
About This Item
Code UNSPSC :
41106300
Nomenclature NACRES :
NB.15
Matériaux:
(with OptiTrack high contrast labelling)
polypropylene
Volume des puits:
150 μL
Stérilité:
non-sterile
Caractéristiques:
lid: no
skirt
165,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
(with OptiTrack high contrast labelling)
polypropylene
Stérilité
non-sterile
Caractéristiques
lid: no
skirt
Conditionnement
pkg of 25 ea
Fabricant/nom de marque
Eppendorf® 0030129857
Caractéristiques du produit alternatif plus écologique
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
Volume des puits
150 μL
Adéquation
suitable for PCR
Autre catégorie plus écologique
Description générale
Eppendorf® twin.tec PCR Plate 96, BioBased, skirted, PCR clean, spring green, 25 plates
Caractéristiques et avantages
  • Reducing your environmental footprint inthe lab
  • Produced from second-generation biobased with 90% of thefeedstock attributed to renewableresources (i.e. waste and residues fromvegetable oil refining or waste cooking oil), avoiding the use of fossilfuel-based petroleum
  • Eppendorf production facility is ISCC PLUS (InternationalSustainability & Carbon Certification) certified, confirming the rawmaterials used at the facility are “bio-circular”. Enables traceability alongthe entire supply chain
  • Laser-engraved lot number and expiration date on each single plate to improve traceability in your laboratory processes
  • Unique laser-engraved optical guiding grid and OptiTrack matrix for quick orientation when pipetting manually
  • One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performanceExtremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample
  • Exceptionally solid, torque- and heat-resistant polycarbonate frame: No warping, bending or breaking during heat incubation or centrifugation
  • Raised well rims for effective sealing, also reduces risk of cross-contaminationBatch-tested and certified free of DNA, DNAase, RNase and PCR inhibitors (PCR clean)
Informations légales
Eppendorf is a registered trademark of Eppendorf AG