Principaux documents
E4278
Sartorius eLINE® single channel electronic pipettor
volume 5-120 μL, AC/DC input 110 V / 230 V AC, universal plug set
Synonyme(s) :
Biohit pipettes, electronic pipettes, pipettor
About This Item
Code UNSPSC :
41121500
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Caractéristiques
CE compliant
electronic
single-channel
Fabricant/nom de marque
Sartorius 730041
Volume
5-120 μL
Description générale
Sartorius eLINE electronic pipettors of the Biohitfamily offer the latest technology in liquid handling. The eLINE electronic pipettor has been designed to deliver the highest levels of analytical accuracy with minimal the eLINE pipettors together with the ejection - take the effort out of pipetting in every laboratory.
The eLINE modes including:
  • Forward Pipetting with Optional Cycle
  • Reverse Pipetting
  • Manual Pipetting
  • Post Delivery Mixing
  • Multi-Dispensing
  • Multi-Aspirating
  • Automated Multi-Dispensing
  • Sequential-Dispensing
Supplied with a universal adapter for Euro, UK and China plugs.
Informations légales
eLINE is a registered trademark of Sartorius Biohit Liquid Handling Oy