DWP961000G2-EP
Eppendorf® Deepwell Plate 96/1000μL
PCR Clean, volume 1000 μL, green border with clear wells, pkg of 20 plates (5 bags x 4 plates)
About This Item
Produits recommandés
Stérilité
non-sterile
Caractéristiques
PCR Clean
green border with clear wells
Conditionnement
pkg of 20 plates (5 bags x 4 plates)
Fabricant/nom de marque
Eppendorf® 951032662
Disponibilité
available only in North America
Technique(s)
PCR: suitable
Taille
96 well
Volume
1000 μL
Puits
96
Description générale
Caractéristiques et avantages
- OptiTrack® matrix: up to 30 % faster sample identification and fewer pipetting errors via high-contrast alphanumeric labeling
- RecoverMax® well design: Optimized well geometry for maximum sample recovery and excellent mixing properties
- Minimal remaining volume and high uniformity from well to well
- Raised well rims and smooth surface for reliable closing, even with heat sealing
- Easy and reliable stacking
- g-Safe: exceptional centrifugation stability up to 6,000 × g for faster protocols and improved sample quality
- High-quality polypropylene guarantees high resistance to chemicals, mechanical stress and temperature extremes
- Manufactured without slip agents, plasticizers, and biocides – Materials which could have a negative effect on bioassays
- Autoclavable (121?°C, 20?min)
- Available with barcode. Additional details available at: www.eppendorf.com/barcode