CLS6798610D
Corning® stirrer with digital display
AC/DC input 230 V AC (CEE7-7 plug), plate L × W 10 in. × 10 in.
Synonyme(s) :
magnetic stirrer, magnetic stirrers, stirrers
870,00 €
About This Item
870,00 €
Produits recommandés
Conditionnement
case of 1
Fabricant/nom de marque
Corning 6798-610D
Disponibilité
not available in USA, Canada and EU
Paramètres
60-1150 rpm speed
Plaque/Plateau, L × l
10 in. × 10 in.
Catégories apparentées
Description générale
- Digital LED stir speed display indicates when set speed has been reached
- Microprocessor maintains consistent and repeatable stir speed settings
- Extremely durable and heat resistant Pyroceram glass-ceramic top
- Small footprint and low profile
- Meets UL and cUL standards