Principaux documents
CLS6796610D
Corning® stirrer with digital display
AC/DC input 230 V AC (Supplied with both 3-pin UK plug and 3-pin Euro plug), plate L × W 10 in. × 10 in.
Synonyme(s) :
magnetic stirrer, magnetic stirrers, stirrers
1 EA
850,00 €
850,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
850,00 €
About This Item
Code UNSPSC :
41103803
Nomenclature NACRES :
NB.73
850,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Conditionnement
case of 1
Fabricant/nom de marque
Corning 6796-610D
Disponibilité
not available in USA and Canada
Paramètres
60-1150 rpm speed
Plaque/Plateau, L × l
10 in. × 10 in.
Description générale
  • Digital LED stir speed display indicates when set speed has been reached
  • Microprocessor maintains consistent and repeatable stir speed settings
  • Extremely durable and heat resistant Pyroceram glass-ceramic top
  • Small footprint and low profile
  • Meets UL and cUL standards
Autres remarques
individual units are not routinely calibrated, rpm speeds are approximate only
Informations légales
Corning is a registered trademark of Corning, Inc.