Principaux documents
CLS4880
Corning® Multipette® adjustable volume multichannel pipettor
12-pette
About This Item
Code UNSPSC :
41121500
Nomenclature NACRES :
NB.78
Caractéristiques
12-channel
electronic: no
Fabricant/nom de marque
Corning 4880
Plage de mesure du volume
20-200 μL
Volume
200 μL
Description générale
1-12 channel pipette
The 8-Pette and 12-Pette offer features for ease of use, versatility, and safety:
  • 20-200 μL per channel in 0.5 μL increments
  • Fully autoclavable
  • Pistol grip eliminates thumb fatigue
  • Separate tip ejector prevents accidental ejection
  • Any number of channels can be left un-used without affecting accuracy
  • Horizontal configuration moves hand closer to tips for better control of placement
Informations légales
Corning is a registered trademark of Corning, Inc.
Multipette is a registered trademark of Eppendorf AG
Articles
Take a deep dive into the world of 3D cell culture courtesy of Corning and understand why the Corning® spheroid microplate can be an asset in the evolution of 3D cell culture.
Protocoles
Understand how Corning® Spheroid Microplates can be used for generating and culturing tumor spheroids in a 96-well ­spheroid microplate format.