Principaux documents
CLS440826
Corning® nine-position stirrer integral control
AC/DC input 120 V AC, US 3-pin plug
Synonyme(s) :
Corning stirrer accessory
About This Item
Code UNSPSC :
41103803
Nomenclature NACRES :
NB.73
Le produit CLS440826 n'est pas actuellement commercialisé dans votre pays Contacter notre Service technique
Description générale
This Corning nine position stirrer has integral controls that can accommodate up to nine 250mL vessels. It operates on 120V/60Hz and is UL approved to meet needs in North America and other locations with similar requirements.
  • Compact design saves valuable bench space
  • Offers powerful stirring from 350 to 2000 rpm.
  • Rugged lightweight glass filled resin case
  • Two-year warranty
Informations légales
Corning is a registered trademark of Corning, Inc.