Principaux documents
CLS432074
Corning® ThermalTray
LP medium profile for ice
Synonyme(s) :
lab equipment accessory
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
NB.12
Caractéristiques
LP medium profile for ice
Conditionnement
pkg of 1
Fabricant/nom de marque
Corning 432074
L × l × H
28.0 cm × 14.0 cm × 8.6 cm
Description générale
Corning ThermalTray thermo-conductive platforms support CoolRack and CoolSink sample modules in liquid temperature sources such as melting ice, water baths and liquid nitrogen. Made of the same highly conductive alloy as CoolRack and CoolSink modules, ThermalTray platforms conduct the source temperature to the CoolRack or CoolSink and, ultimately, to your samples.
  • Stable, sturdy design makes them ideal for processing temperature-sensitive samples in melted ice baths or liquid nitrogen.
  • All modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
Informations légales
Corning is a registered trademark of Corning, Inc.
ThermalTray is a trademark of Corning, Inc.