CLS431444
Corning® cap, Erlenmeyer access, LUER, 1 L
Synonyme(s) :
Erlenmeyer flask cap
632,00 €
About This Item
632,00 €
Produits recommandés
Fabricant/nom de marque
Corning 431444
Disponibilité
available only in South Korea
Description générale
- Aseptic transfer caps are chemically resistant and fit Corning® Erlenmeyer 1L flasks.
- Features a venting port that ends in a 50mm diameter vent filter with a 0.2 μm pore Polyvinylidene fluoride (PVDF) membrane and a barbed fitting.
- The liquid transfer port has an internal end with a rigid polypropylene dip tube angled to reach the bottom of the flask.
- Heat sealable flexible tubing ends in a male Luer lock (MLL) closed with a female Luer sealing cap.